Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sensititre™ Anaerobe MIC Plate

Catalog No. STAN02B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STAN02B 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. STAN02B Supplier Thermo Scientific™ Supplier No. YANO2B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform high-quality susceptibility testing for anaerobes and B. fragilis group with the Thermo Scientific™ Sensititre™ Anaerobe MIC Plate.

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

0.5 McFarland Standard MHB (Part No. T346205)
Antimicrobial Agents Various
Broth Type Brucella Broth (Part No. T3450)
Description Sensititre Anaerobe MIC Plate
Antibiotic Code Various
Product Type Anaerobic Plate
Target Organism Class Anaerobes
Test Type Minimum Inhibitory Concentration (MIC)
For Use With (Application) Anaerobes, B. fragilis group
Incubation Conditions 34-36°C O2
Incubation Time 46-48 hr
Isolates Per Plate 1
Quantity 10/Pk.
Reading Options Manual
Transfer Volume 100µL
Volume Per Well 100 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.