Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sensititre™ ARIS HiQ™ System for AST

Catalog No. styv4000
Change view
Click to view available options
Description:
PC Kit
Sensititre ARIS HiQ
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
STYV4000 Sensititre ARIS HiQ
STYSW4000 PC Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. STYV4000 Supplier Thermo Scientific™ Supplier No. YV4000

This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

This item is not returnable. View return policy

Achieve comprehensive, accurate MIC results to guide targeted patient therapy with this benchtop, automated reading and incubation system for AST.

Achieve the comprehensive, accurate MIC values needed to guide more targeted patient therapy with the Thermo Scientific™ Sensititre™ ARIS HiQ™ System, a benchtop automated incubation and reading system for antimicrobial susceptibility testing and identification of bacterial pathogens; designed to accommodate up to 100 Sensititre™ microtitre plates within a single instrument.

Confidently detect emerging antibiotic resistance with the gold-standard level accuracy1,2 of broth microdilution coupled with time-saving benefits of automation to optimize patient care, support antimicrobial stewardship, and improve lab efficiency with the Sensititre™ ARIS HiQ™ System.   

  • Efficient plate handling: central turntable for loading and unloading Sensititre plates via robotic plate transfer and advanced sorting options to optimize read times and efficient plate removal
  • Flexible loading: batch load and unload capability for improved testing workflow
  • Seamless LIS connectivity: Thermo Scientific™ Sensititre SWIN™ software connection is configured to meet LIS connectivity requirements
  • Large capacity, small footprint: to process more tests at one time with limited impact to bench space.

 

The Sensititre ARIS HiQ system helps microbiology laboratories deliver accurate antimicrobial susceptibility testing of the most commonly prescribed antibiotics, as well as novel, last-resort therapies, to provide meaningful results for clinicians, stewardship teams, and public health departments. By providing critical data to support decisions on prescribing the right antibiotic, at the right dose, at the right time, the Sensititre AST System supports the effort to improve patient outcomes and to ensure the right actions are taken to protect the future of healthcare, veterinary, and agriculture industries against the threat of antibiotic resistance.

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 0°C to 40°C (32°F to 104°F)
Description Sensititre ARIS HiQ
Product Type Microbiology Media
Test Type Minimum Inhibitory Concentration (MIC)
Width (English) 29.53 in.
Width (Metric) 750 mm
Height (English) 44.31 in.
Height (Metric) 1125 mm
Depth (English) 28.94 in.
Depth (Metric) 735 mm
Weight (English) System, empty (no racks, no plates): 90KG (198.5lb)
Weight (Metric) System, full (11 racks - 10 internal racks, 1 utility rack, 100 plates,100μL water per plate): 103KG (226.6lb)
Show More Show Less

Thermo Fisher Scientific products are distributed globally so uses, applications, and availability of product in each country depend on local regulatory marketing authorization status.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.