Learn More
Thermo Scientific™ Sensititre™ Complete Automated AST System
Catalog No. YV3000VZ Shop All Thermo Scientific Products
Change view
Click to view available options
Test Type:
Minimum Inhibitory Concentration (MIC)
Catalog No. Test Type
YV3000VZ Minimum Inhibitory Concentration (MIC)
1 options
This item is not returnable. View return policy
Catalog No. YV3000VZ Supplier Thermo Scientific™ Supplier No. YV3000VZ
Request a Quote
This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
This item is not returnable. View return policy
Perform all susceptibility testing on a single platform utilizing the superior sensitivity of true MIC results with the Thermo Scientific™ Sensititre™ Complete Automated AST System.
Perform all susceptibility testing on a single platform utilizing the superior sensitivity of true MIC results with the Thermo Scientific™ Sensititre™ Complete Automated AST System. The complete automated system includes the Sensititre™ ARIS™ HiQ™, OptiRead™, Sensititre AIM™, Sensititre™ Vizion™ System, Sensititre™ SWIN™ computer, Nephelometer, barcode printer and barcode labels.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Description Sensititre Complete Automated AST System
Product Type Sensititre Complete Automated System
Test Type Minimum Inhibitory Concentration (MIC)