Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sensititre™ Equine EQUIN1F Vet AST Plate

Catalog No. STYEQUIN1F Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STYEQUIN1F 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. STYEQUIN1F Supplier Thermo Scientific™ Supplier No. EQUIN1F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Perform accurate AST with an equine-specific formulary with this convenient plate.

Perform accurate AST with an equine-specific formulary with the Thermo Scientific™ Sensititre™ Equine EQUIN1F AST Plate.

Achieve reliable, consistent results for non-fastidious Gram-positive and Gram-negative isolates of veterinary origin with Sensititre Equine EQUIN1F AST Plate.

  • Extended shelf life, room temperature storage eliminates inventory control concerns.
  • Individual packaging allows the laboratory to test one plate at a time with no waste.
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology.
  • Automated or manual read options are available for ultimate flexibility.

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Various
Description Sensititre Equine MIC Plate
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Non-fastidious, Gram-positive and Gram-negative Veterinary Isolates
Test Type Veterinary AST, Minimum Inhibitory Concentration (MIC)
Isolates Per Plate 1
Quantity 10/Pk.
Reading Options Autoread or manual
Specimen Type Equine Isolates
Species Equine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.