Learn More
Description
Confidently perform susceptibility testing of Gram-negative isolates on a single plate which includes antimicrobials ceftazidime/avibactam and ceftolozane/tazobactam using Thermo Scientific™ Sensititre™ Gram Negative GN7F AST Plates.
Address your formulary needs with this FDA-cleared, micro-broth dilution plate for AST which meets CLSI standards.
Obtain true minimum inhibitory concentration (MIC) results to both inform targeted therapy decisions for resistant infections with otherwise limited or no treatment options and to support antimicrobial stewardship to preserve the effectiveness.
Expand treatment options by performing ceftazidime/avibactam and ceftolozane/tazobactam susceptibility testing of Gram-negative organisms with Sensititre Gram Negative GN7F AST Plates. Generate clinically relevant data, detect emerging resistance, and help guide timely and accurate therapy intervention by utilizing micro-broth dilution.
- Accurate: meets CLSI and FDA breakpoints
- Quantitative: precise MIC results aid informed patient therapy decisions
- Efficient: accurate results the first time, thereby reducing unnecessary testing and protocols
- Flexible: manual to automated read options are available for ultimate flexibility
- Easy-to-store: extended shelf life and room-temperature storage simplify inventory control
The Sensititre System is a leader in AST solutions, offering flexible, customizable testing options for laboratories of all sizes.
- Scalable: flexible instrumentation options designed to be modular in order to meet the unique needs of individual lab space, workload, and budget requirements
- Flexible: over 300 antimicrobials available in extended dilution ranges, access to the newest antimicrobials, and the ability to consolidate AST onto one platform
- Tailor-made: design your own AST panel to meet specific formulary requirements and prescription protocols and to clinicians that are based on real-time trends and local epidemiology.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| 0.5 McFarland Standard | Demineralized Water (part no. T3339) |
| Antimicrobial Agents | Various |
| Broth Type | MHB (part no. T3462) |
| Description | Sensititre Gram Negative GN7F AST Plate |
| Antibiotic Code | Various |
| Product Type | AST Plate |
| Target Organism Class | Non-fastidious Gram-negative Organisms |
| Test Type | Minimum Inhibitory Concentration (MIC) |
| Incubation Conditions | 35°C; O2 |
| Incubation Time | 18-24 hrs |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.