Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sensititre™ Gram Negative MDRGN4F AST Plate, IVD

Catalog No. 23900177 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-900-177 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-900-177 Supplier Thermo Scientific™ Supplier No. YMDRGN4F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Expand your Gram-negative organism susceptibility testing options by accessing the latest antibiotics including sulbactam-durlobactam. The Thermo Scientific™ Sensititre™ MDRGN4F plate delivers results directly from broth microdilution (BMD), the gold standard1 phenotypic method for antimicrobial susceptibility testing (AST).

Expand your Gram-negative organism susceptibility testing options by accessing the latest antibiotics including sulbactam-durlobactam. The Thermo Scientific™ Sensititre™ MDRGN4F plate delivers results directly from broth microdilution (BMD), the gold standard1 phenotypic method for antimicrobial susceptibility testing (AST). Open access to one of the most comprehensive ranges of second-line antimicrobials, on a single plate format, designed for in-vitro diagnostic use.

Treating multidrug-resistant infections and monitoring emergent multidrug resistance is more important now than ever. Our Thermo Scientific™ Sensititre™ plates provide clinicians with gold standard-level, accurate minimum inhibitory concentration (MIC) results to guide optimal treatment decisions when it matters.

  • Updated standard format including sulbactam-durlobactam
  • IVD-labelled
  • BMD provides accurate MIC results therefore negating the need for repeat testing 
  • Designed for use with CLSI breakpoints
  • Sensititre MIC determination is achieved using direct measurement rather than relying on extrapolated data, thereby providing an accurate MIC result, not an estimate. 
  • All contained antibiotics also available in customizable plate options, along with access to over 300 antimicrobials, to enable meeting local formulary and stewardship program requirements

Not all products are available for sale in all territories. Please inquire.

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Various
Broth Type Mueller Hinton Broth (T3462)
Description Thermo Scientific™ Sensititre™ Gram Negative IVD AST Plate
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Non-fastidious Gram-negative Organisms
Quantity 10/Pk.
Reading Options Manual or Automated
Volume Per Well Reconstitution volume - 50µL
Species Non-fastidious Gram-negative Isolates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.