Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sensititre™ Gram Negative MIC Plate

Catalog No. STGN3F Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
MIC Plate
MIC plate for CRE testing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
STGN3F MIC plate for CRE testing
STGN2F MIC Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. STGN3F Supplier Thermo Scientific™ Supplier No. GN3F

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Utilize the latest antimicrobials, including Doripenem and Tigecycline, for accurate, economical results with Thermo Scientific™ Sensititre™ Gram Negative MIC Plates.

Utilize the latest antimicrobials, including Doripenem and Tigecycline, for accurate, economical results with Sensititre Gram Negative MIC Plates.

  • Multi-isolate format decreases cost per test
  • Automated, semiautomated or manual read options are available for ultimate flexibility
  • Extended shelf life and room-temperature storage simplify inventory control

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

0.5 McFarland Standard H2O (Part No. T3339)
Antimicrobial Agents Various
Broth Type MHB (Part No. T3462)
Description MIC plate for CRE testing
Antibiotic Code Various
Product Type MIC Plate
Target Organism Class Gram-negative Bacteria
Test Type Minimum Inhibitory Concentration (MIC), Carbapenem-resistant Enterobacterales (CRE)
For Use With (Application) Gram negative organisms: Enterobacteriaceae, Non-Enterobacteriaceae, Proteus sp.
Incubation Conditions 35°C O2
Incubation Time 18 hr
Isolates Per Plate 1
No. per Pack 10
Packaging Type Box
Quantity 10/Pk.
Reading Options Autoread or manual
Transfer Volume For Enterobacteriaceae and Non-Enterobacteriaceae: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp.: 1μL
Volume Per Well 50 μL
Species Multiple
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.