Learn More
Thermo Scientific™ Sensititre™ Gram Negative Non-Fermenters MIC Plate
Catalog No. STNF Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
STNF 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. STNF Supplier Thermo Scientific™ Supplier No. YNF
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Obtain accurate, reliable MIC results for non-fermenters isolates with the Thermo Scientific™ Sensititre™ Gram Negative Non-Fermenters MIC Plate.
Obtain accurate, reliable MIC results for non-fermenters isolates with the Sensititre Gram Negative Non-Fermenters MIC Plate.
  • Multi-isolate format decreases cost per test
  • Automated, semiautomated or manual read options are available for ultimate flexibility
  • Extended shelf life and room-temperature storage simplify inventory control
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
0.5 McFarland Standard H2O (Part No. T3339)
Antimicrobial Agents Various
AST Type MIC
Broth Type MHB (Part No. T3462)
Description Sensititre Gram Negative Non-Fermenters MIC Plate
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Gram-negative, Non-fermenting Organisms
Test Type Minimum Inhibitory Concentration (MIC)
Incubation Conditions 35°C O2
Incubation Time 18 hr
Isolates Per Plate 1
No. per Pack 10
Quantity 10/Pk.
Reading Options Autoread or manual
Transfer Volume For Enterobacteriaceae and Non-Enterobacteriaceae: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp.: 1μL
Volume Per Well 50 μL