Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sensititre™ Gram Positive GPALL3F AST Plate

Catalog No. STGPALL3F Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STGPALL3F 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. STGPALL3F Supplier Thermo Scientific™ Supplier No. GPALL3F

missing translation for 'validMainframeMsg'

Simplify testing of non-fastidious Gram-positive isolates with this plate containing CLSI breakpoint ranges for Ceftaroline and Telavancin

Perform susceptibility testing of non-fastidious Gram-positive isolates on a single platform with Thermo Scientific™ Sensititre™ Gram Positive GPALL3F AST Plate. The convenient standard AST plate includes updated CLSI breakpoints for Ceftaroline and Telavancin.

Achieve reliable, consistent results for non-fastidious Gram-positive isolates with the Sensititre Gram Positive GPALL3F AST Plate. Generate clinically relevant data, detect emerging resistance, and help guide timely and accurate therapy intervention by utilizing micro-broth dilution.

  • Plate has fewer limitations for Enterococcus and Staphylococcus spp. versus other automated systems
  • Full-range, true MIC readings provide results for better detection of resistance
  • Automated or manual read options are available for ultimate flexibility
  • Extended 18 to 24 month shelf life and room-temperature storage simplify inventory control

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

0.5 McFarland Standard H2O (Part No. T3339)
Antimicrobial Agents Various
Broth Type MHB (Part No. T3462)
Description Sensititre Gram Positive All-in-One Plate, with Ceftaroline and Telavancin
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Non-fastidious Gram-positive Organisms
Test Type Minimum Inhibitory Concentration (MIC)
Incubation Conditions 35°C O2
Incubation Time 18-24 hr
Isolates Per Plate 1
No. per Pack 10
Quantity 10/Pk.
Reading Options Autoread or manual
Transfer Volume 10µL or 30µL* (*Aid in detection of resistance mechanisms for gram positive organisms; see pack insert)
Volume Per Well 50 μL
Species Non-fastidious Gram-positive Isolates
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.