Learn More
Thermo Scientific™ Sensititre™ Gram Positive GPALL3F AST Plate
Catalog No. STYGPALL3F Shop All Thermo Scientific Products
Change view
Click to view available options
Volume Per Well:
50 μL
Catalog No. Volume Per Well
STYGPALL3F 50 μL
1 options
This item is not returnable. View return policy
Catalog No. STYGPALL3F Supplier Thermo Scientific™ Supplier No. YGPALL3F
Add to Cart
Add to Cart
This item is not returnable. View return policy
Simplify testing of non-fastidious Gram-positive isolates with this plate containing CLSI breakpoint ranges for Ceftaroline and Telavancin.
Achieve reliable, consistent results for non-fastidious Gram-positive isolates with the Sensititre Gram Positive GPALL3F AST Plate. Generate clinically relevant data, detect emerging resistance, and help guide timely and accurate therapy intervention by utilizing micro-broth dilution.
  • Plate has fewer limitations for Enterococcus and Staphylococcus spp. versus other automated systems
  • Full-range, true MIC readings provide results for better detection of resistance
  • Automated or manual read options are available for ultimate flexibility
  • Extended 18 to 24 month shelf life and room-temperature storage simplify inventory control
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
0.5 McFarland Standard H2O (Part No. T3339)
Broth Type MHB (Part No. T3462)
Product Type AST Plate
Target Organism Class Non-fastidious, Gram-negative Organisms
Test Type Minimum Inhibitory Concentration (MIC)
Incubation Conditions 35°C O2
Incubation Time 18 to 24 hr.
Isolates Per Plate 1
No. per Pack 10
Quantity 10/Pk.
Reading Options Autoread or Manual
Transfer Volume 10 or 30 μL (Aid in detection of resistance mechanisms for gram positive organisms; see pack insert)
Volume Per Well 50 μL