Learn More
Thermo Scientific™ Sensititre™ HPB1
Catalog No. STHPB1 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
STHPB1 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. STHPB1 Supplier Thermo Scientific™ Supplier No. HPB1
This item is not returnable. View return policy
Confidently perform susceptibility testing of Streptococcus pneumoniae and Haemophilus influenza with this FDA cleared, convenient plate.
Perform high-quality susceptibility testing with true MIC results using the Thermo Scientific™ Sensititre™ Haemophilus and Streptococcus HPB1 AST Plate. Address your formulary needs with this FDA-cleared, micro-broth dilution plate for antimicrobial susceptibility testing.
Achieve reliable, consistent results for Streptococcus pneumoniae and Haemophilus influenzae isolates with Sensititre Haemophilus and Streptococcus HPB1 AST Plate
  • Quantitative: precise MIC results aid informed patient therapy decisions
  • Efficient: accurate results the first time, thereby reducing unnecessary testing and protocols
  • Easy-to-store: extended shelf life and room-temperature storage simplify inventory control
  • Economic: individual packaging allows laboratory to test one plate at a time with no waste
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
0.5 McFarland Standard MHB (Part No. T346205)
Antimicrobial Agents Various
Broth Type For Haemophilus: HTM (Part No. T3470). For S. pneumoniae/Streptococcus sp.: MHB w/ LHB (Part No. CP112)
Description Sensititre Haemophilus and Streptococcus pneumoniae MIC Plate
Antibiotic Code Various
Product Type AST Plate
Test Type Minimum Inhibitory Concentration (MIC)
Incubation Conditions 34-36°C O2
Incubation Time 20-24 hr
Isolates Per Plate 1
No. per Pack 10
Quantity 10/Pk.
Reading Options Manual
Transfer Volume For Haemophilus: 50μL. For S. pneumoniae/Streptococcus sp.: 100μL
Volume Per Well 100 μL
Species Streptococcus pneumoniae, Haemophilus influenzae