Description
Perform simple visual reads of your 96-well microtitre plates with the mirrored Thermo Scientific™ Sensititre™ Manual Viewbox.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Description | Sensititre Manual Viewbox |
| Product Type | Manual Viewbox |
| Test Type | Minimum Inhibitory Concentration (MIC) |