Learn More
Thermo Scientific™ Sensititre™ Gram-Negative MDRGNX4F AST Plate RUO
Shop All Thermo Scientific ProductsDescription
Expand your Gram-negative organism susceptibility testing options by accessing the latest antibiotics including sulbactam-durlobactam. The Thermo Scientific™ Sensititre™ MDRGNX4F Plate delivers results directly from broth microdilution (BMD), the gold standard1 phenotypic method for Antimicrobial Susceptibility Testing (AST). Open access to one of the most comprehensive ranges of second-line antimicrobials, together with colistin and fosfomycin on a single plate format, designed for research use only.
Treating multidrug-resistant infections and monitoring emergent multidrug resistance is more important now than ever. Our Thermo Scientific™ Sensititre™ Plates provide clinicians with gold standard-level, accurate minimum inhibitory concentration (MIC) results to guide optimal treatment decisions when it matters.
- Updated standard format including sulbactam-durlobactam, imipenem/relebactam, colistin and fosfomycin
- Research Use Only (RUO)
- BMD provides accurate MIC results therefore negating the need for repeat testing
- Designed for use with CLSI breakpoints
- Sensititre MIC determination is achieved using direct measurement rather than relying on extrapolated data, thereby providing an accurate MIC result, not an estimate.
- All contained antibiotics are also available in customizable plate options, along with access to over 300 antimicrobials, to enable the meeting of local formulary and stewardship program requirements
Not all products are available for sale in all territories. Please inquire.
Specifications
Specifications
| Antimicrobial Agents | Various |
| Broth Type | Demineralized Water (T3339), Mueller Hinton Broth (T3462) |
| Description | Sensititre™ Gram-Negative RUO AST Plate |
| Antibiotic Code | Various |
| Product Type | AST Plate |
| Target Organism Class | Non-fastidious Gram-negative Organisms |
| For Use With (Application) | Susceptibility testing of non-fastidious, gram negative isolate |
| Quantity | 10/Pk. |
| Reading Options | Manual or Automated |
| Volume Per Well | Reconstitution Volume - 50 µL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.