Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sensititre™ Nephelometer

Catalog No. STYV3011 Shop All Thermo Scientific Products
Change view
Click to view available options
Weight (Metric):
0.8 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
STYV3011 0.8 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. STYV3011 Supplier Thermo Scientific™ Supplier No. YV3011
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cost-effectively standardize inoculum density with the Thermo Scientific™ Sensititre™ Nephelometer.

Optimize performance and achieve accurate results by standardizing bacterial suspension density with the Thermo Scientific™ Sensititre™ Nephelometer. Designed to rapidly prepare a consistent inoculum density, the easy-to-read LED indicator light offers a quick methodology to produce bacterial suspensions equivalent to the required 0.5 McFarland Standard.

The Sensititre Nephelometer reduces the risk of manual errors associated with targeting numeric values, by guiding the user to increase/decrease the concentration of emulsified test colonies, until a green “in range” result is achieved – indicating that the suspension is ready for addition to the chosen test broth.

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Description Sensititre Nephelometer
Product Type Nephelometer
Test Type Minimum Inhibitory Concentration (MIC)
Width (English) 7.5 in.
Width (Metric) 19 cm
Height (English) 3.84 in.
Height (Metric) 9.7 cm
Depth (English) 5.3 in.
Depth (Metric) 13.5 cm
Weight (English) 1.8 lb.
Weight (Metric) 0.8 kg
Electrical Requirements 8-18VAC/DC; 130mA; 1w
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.