Learn More
Description
Perform susceptibility testing of Streptococcus species with this FDA cleared, convenient plate including Tigecycline.
Perform susceptibility testing of Streptococcus species with Thermo Scientific™ Sensititre™ Streptococcus STP6F AST Plate. This plate is FDA-cleared for automated or manual reading of Streptococcus species using the Thermo Scientific™ Sensititre™ ARIS™ 2X ID/AST System, Thermo Scientific™ Sensititre™ OptiRead System, or Sensititre™ manual platforms, including the Thermo Scientific™ Sensititre™ Vizion™ System for maximum versatility.
Eliminate offline tests with a full range of antimicrobials, including Tigecycline, with the Sensititre Streptococcus STP6F AST Plate.
- Individual packaging allows laboratories to test one isolate at a time
- Test 20 antimicrobials on a single AST plate for cost-effective results
- True MIC results provide better resistance tracking
- Extended shelf life and room-temperature storage eliminate inventory control concerns
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| 0.5 McFarland Standard | MHB (Part No. T346205) |
| Antimicrobial Agents | Various |
| Broth Type | MHB w/ LHB (Part No. CP114) |
| Description | Sensititre Streptococcus species MIC Plate |
| Antibiotic Code | Various |
| Product Type | AST Plate |
| Target Organism Class | Fastidious Organisms |
| Test Type | Minimum Inhibitory Concentration (MIC) |
| For Use With (Application) | All Streptococcus sp. |
| Incubation Conditions | 34-36°C O2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.