Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sensititre™ Streptococcus STP8F AST Plate

Catalog No. STYSTP8F Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STYSTP8F 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. STYSTP8F Supplier Thermo Scientific™ Supplier No. YSTP8F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform susceptibility testing of Streptococcus species with Thermo Scientific™ Sensititre™ Streptococcus STP8F AST Plate.

Perform susceptibility testing of Streptococcus species with the Thermo Scientific™ Sensititre™ Streptococcus STP8F AST Plate. This plate design follows CLSI and FDA breakpoints for Streptococcus species and can be read automatically using the Thermo Scientific™ Sensititre™ ARIS HiQ™ System, Thermo Scientific™ Sensititre™ OptiRead™ System, or Sensititre™ manual platforms, including the Thermo Scientific™ Sensititre™ Vizion™ System for maximum versatility.

Eliminate offline tests with a full range of antimicrobials using the Sensititre Streptococcus STP8F AST Plate.

  • Individual packaging allows laboratories to test one isolate at a time
  • Test 21 antimicrobials on a single AST plate for cost-effective results
  • True MIC results provide better resistance tracking
  • Room-temperature storage throughout the shelf life eliminates inventory control concerns


Not all products are available for sale in all territories. Please inquire.

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Various
Broth Type MHB w/ LHB (Part No. CP11410)
Description Sensititre Streptococcus species MIC Plate
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Fastidious Organisms
Quantity 10/Pk.
Reading Options Manual or Automated
Transfer Volume 100 µL
Volume Per Well 100 µL
Species Streptococcus species
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.