Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sensititre™ SWIN™ Software System

Catalog No. p-4525214
Change view
Click to view available options
Description:
22 in. touch screen monitor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
STSW1302 22 in. touch screen monitor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. STSW1302 Supplier Thermo Scientific™ Supplier No. SW1302

Item Discontinued This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

Consolidate your entire test program on a single software platform while enhancing data entry and reporting with the Thermo Scientific™ Sensititre™ SWIN™ Software System.

Consolidate your entire test program on a single software platform while enhancing data entry and reporting with the Thermo Scientific™ Sensititre™ SWIN™ Software System. Simple touch screen navigation puts results at your fingertips while drop-down fields eliminate manual entry errors for all manual, semi-automated and fully automated reads.

Consolidate your entire test program on a single software platform while enhancing data entry and reporting with the Sensititre SWIN Software System.
  • Customizable Expert System provides three-tiered expert messages: Information, Modification (based on FDA, CLSI and EUCAST recommendations) and Warning Levels
  • Quality control module easily manages MIC results and broth information for inspection purposes
  • Automated alerts and report functionality enhance workflow and expedite release of results

Includes:
2 GB of RAM, 2-80 GB SATA Hard Drives, DVD RW Drive, 6+ USB ports, 1 Parallel port, keyboard, mouse, internal modem, barcode scanner, Laser Jet printer, Windows XP Professional, 8-port Serial to USB Converter, PC Anywhere, 17" Touch screen Monitor, SWIN Software

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Description 22 in. touch screen monitor
Product Type Microbiology Media
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.