Learn More
Thermo Scientific™ Sensititre™ YeastOne™ YO3IVD AST Plate
Catalog No. STYO3IVD Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
STYO3IVD 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. STYO3IVD Supplier Thermo Scientific™ Supplier No. YYO3IVD
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Obtain accurate results for antimicrobial susceptibility of invasive Candida infections to guide therapy with this convenient plate containing Micafungin
Obtain accurate results for antifungal susceptibility of invasive Candida infections to guide therapy with Thermo Scientific™ Sensititre™ YeastOne™ YO3IVD AST Plate with Micafungin. Report and track echinocandin susceptibility against Candida spp. and make informed decisions to step-down patient therapy with this FDA-cleared panel which includes micafungin.
Simplify and consolidate antifungal susceptibility testing on a single plate with Sensititre YeastOne YO3IVD AST Plate with Micafungin. Colorimetric Thermo Scientific™ alamarBlue™ agent provides reliable, easy, and consistent endpoint determination with visual read option for efficient workflow and accurate results.
  • Wide range of FDA-cleared antifungal compounds on a single susceptibility plate simplifies workflows
  • Complete echinocandin information, with the addition of micafungin, supports susceptibility tracking and reporting
  • Broad dilution ranges allow earlier detection of susceptibility changes and emerging resistance
  • Room-temperature storage eliminates inventory control concerns
  • Individual packaging allows the laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • 24-hour incubation time enables timely results
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand
TRUSTED_SUSTAINABILITY
Specifications
0.5 McFarland Standard MHB (Part No. T346205)
Antimicrobial Agents Various
Broth Type For YeastOne Broth: (Part No. Y3462)
Description Sensititre YeastOne IVD Plate, with Micafungin
Antibiotic Code Various
Product Type Microbiology Media
Test Type Yeast Susceptibility
Incubation Conditions 35°C Non-CO2
Incubation Time 24 hr
Isolates Per Plate 1
Quantity 10/Pk.
Reading Options Manual
Specimen Type Candida Species
Transfer Volume 20 µL
Volume Per Well 100 μL