Description
Simplify yeast susceptibility testing with this plate.
Simplify yeast susceptibility testing with the Thermo Scientific™ Sensititre™ YeastOne™ YO11 AST Plate including Micafungin and Rezafungin. The Sensititre YeastOne YO11 AST Plate is intended for research use only. Colorimetric Thermo Scientific™ alamarBlue™ agent provides reliable, easy and consistent endpoint determination with visual read option for efficient workflow and accurate results.
Achieve reliable, consistent results for Candida species with the Sensititre YeastOne YO11 AST Plate.
- Room-temperature storage eliminates inventory control concerns
- Individual packaging allows laboratory to test one plate at a time with no waste
- Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
- 24-hour incubation time enables timely results for clinicians
Specifications
Specifications
| 0.5 McFarland Standard | H2O (Part No. T3339) |
| Antimicrobial Agents | Various |
| Broth Type | YeastOne Broth (Part No. Y3462) |
| Description | Sensititre YeastOne Research Use Only Plate, w/ Micafungin and Rezafungin |
| Antibiotic Code | Various |
| Product Type | AST Plate |
| Target Organism Class | Yeast |
| Test Type | Yeast Susceptibility |
| Incubation Conditions | 35°C non-CO2 |
| Incubation Time | 24-25 hr or 48-72 hr for Cryptococcus* (*For research use only. Not for use in diagnostic procedures.) |
| Show More |