Description
Obtain accurate results for antifungal susceptibility of invasive Candida infections to guide therapy with this plate.
Obtain accurate results for antifungal susceptibility of invasive Candida infections to guide therapy with Thermo Scientific™ Sensititre™ YeastOne™ YO4IVD AST Plate with Rezafungin. Report and track echinocandin susceptibility against Candida spp. and make informed decisions to step-down patient therapy with this FDA-cleared panel.
Simplify and consolidate antifungal susceptibility testing on a single plate with Sensititre YeastOne YO4IVD AST Plate with Rezafungin. Colorimetric Thermo Scientific™ alamarBlue™ agent provides reliable, easy, and consistent endpoint determination with visual read option for efficient workflow and accurate results.
- Wide range of FDA-cleared antifungal compounds on a single susceptibility plate simplifies workflows
- Complete echinocandin information, with the addition of micafungin, supports susceptibility tracking and reporting
- Broad dilution ranges allow earlier detection of susceptibility changes and emerging resistance
- Room-temperature storage eliminates inventory control concerns
- Individual packaging allows the laboratory to test one plate at a time with no waste
- Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
- 24-hour incubation time enables timely results
Specifications
Specifications
| 0.5 McFarland Standard | 0.5 McFarland Standard (E1041) Demineralized Water (T3339) |
| Antimicrobial Agents | Various |
| Broth Type | For YeastOne Broth: (Part No. Y3462) |
| Description | Sensititre YeastOne IVD Plate, with Rezafungin |
| Antibiotic Code | Various |
| Product Type | AST Plate |
| Target Organism Class | Yeast |
| Test Type | Yeast Susceptibility |
| Incubation Conditions | 35°C Non-CO2 |
| Incubation Time | 24 hr |
| Show More |