Learn More
Bachem HIV Protease Substrate VI
SDP
Supplier:  Bachem 4030763.0001
Encompass_Preferred
HIV Protease Substrate VI, H-Arg-Val-Nle-p-nitro-Phe-Glu-Ala-Nle-NH2, 130877-92-8, C40H66N12O11, Mr 891.04, 1mg. Sensitive chromogenic substrate for both the HIV-1 and HIV-2 proteases.
Catalog No. 50-194-6952