Learn More
Bachem H-Gly-Arg-AMC
SDP
Supplier:  Bachem 4002196.0250
Encompass_Preferred
H-Gly-Arg-AMC Hydrochloride, 65147-19-5, C18H24N6O4, Mr 388.43, 250mg. Sensitive fluorogenic substrate for dipeptidyl aminopeptidase I (DPP I, cathepsin C).
Catalog No. 50-194-7032