Learn More
Bachem Glutaryl-Phe-AMC
SDP
Supplier:  Bachem 4008166.0250
Encompass_Preferred
Glutaryl-Phe-AMC, 58632-47-6, C24H24N2O6, Mr 436.46, 250mg. Sensitive fluorogenic substrate for chymotrypsin.
Catalog No. 50-194-7029