Learn More
AnaSpec SENSOLYTE 520 IDE ACTIVITY KIT
Catalog No. NC0729287
Encompass
SensoLyte 520 IDE Activity Assay Kit Fluorimetric, (1 kit) Insulin degrading enzyme (IDE) is considered a physiological and pathological relevant enzyme. Growing evidence has indicated that IDE may be involved in the pathogenesis of Alzheimer’s disease and diabetes mellitus type 2. The SensoLyte 520 IDE Activity Assay Kit can be used to detect enzyme activity in biological samples or in purified enzyme preparations. The unique FRET substrate was derived from an APP sequence designed to reduce cross reactivity with Neprilysin, ADAM10, TACE, BACE-1, and BACE-2. When active IDE cleaves the FRET substrate, it results in an increase of 5-FAM fluorescence, which is monitored at excitation/emission = 490 nm/520 nm.
Catalog No. NC0729287 Supplier AnaSpec Supplier No. AS72231
SensoLyte 520 IDE Activity Assay Kit Fluorimetric, (1 kit) Insulin degrading enzyme (IDE) is considered a physiological and pathological relevant enzyme. Growing evidence has indicated that IDE may be involved in the pathogenesis of Alzheimer’s disease and diabetes mellitus type 2. The SensoLyte 520 IDE Activity Assay Kit can be used to detect enzyme activity in biological samples or in purified enzyme preparations. The unique FRET substrate was derived from an APP sequence designed to reduce cross reactivity with Neprilysin, ADAM10, TACE, BACE-1, and BACE-2. When active IDE cleaves the FRET substrate, it results in an increase of 5-FAM fluorescence, which is monitored at excitation/emission = 490 nm/520 nm.