Learn More
AnaSpec Green PIN1 Activity Assay Kit Fluorimetric - 1 kit.
Catalog No. NC2282966
Encompass
Green PIN1 Activity Assay Kit Fluorimetric - 1 kit.Component A: Pin1 Green Substrate: 0.1 mM , 60 ¿L Component B: Pin1, Human Recombinant: 1 mg/ml, 10 ¿L Component C: Pin1 Developer, 100X: 50 ¿L, 4 vials Component D: Assay Buffer: 30 mL Component E Pin1 Inhibitor 50mM in DMSO, 25 ¿L
This item is not returnable. View return policy
Catalog No. NC2282966 Supplier AnaSpec Supplier No. AS72240
Green PIN1 Activity Assay Kit Fluorimetric - 1 kit.Component A: Pin1 Green Substrate: 0.1 mM , 60 ¿L Component B: Pin1, Human Recombinant: 1 mg/ml, 10 ¿L Component C: Pin1 Developer, 100X: 50 ¿L, 4 vials Component D: Assay Buffer: 30 mL Component E Pin1 Inhibitor 50mM in DMSO, 25 ¿L