Learn More
AnaSpec SENSOLYTE MG PHOSPHATE ASSAY
Catalog No. NC2069871
Encompass
SensoLyte® MG Phosphate Assay Kit Colorimetric -1 kit. Component A: 1 mM Phosphate, Standard: 1 mL Component B: MG Reagent: 20 mL
Catalog No. NC2069871 Supplier AnaSpec Supplier No. AS71103
SensoLyte® MG Phosphate Assay Kit Colorimetric -1 kit. Component A: 1 mM Phosphate, Standard: 1 mL Component B: MG Reagent: 20 mL