Learn More
AnaSpec SENSOLYTE PNPP ALK ASSY KIT
Supplier:  AnaSpec AS72146
Encompass
SensoLyte pNPP Alkaline Phosphatase Assay Kit *Colorimetric*, (1 kit) The change of alkaline phosphatase activity is involved in a variety of physiological and pathological events, such as bone development, bone-related diseases, gestation-related diseases and inflammatory bowel diseases. Alkaline phosphatase is also a popular enzyme conjugated with secondary antibody for ELISA. p-Nitrophenyl phosphate (pNPP) is proven to be an effective chromogenic substrate for alkaline phosphate. This SensoLyte kit can be used to detect alkaline phosphatase activity in biological samples and alkaline phosphatase-based ELISA. The kit contains: pNPP chromogenic liquid substrate (absorption maximum = 405 nm upon dephosphorylation), Assay buffer, Stop solution, Alkaline phosphatase standard, “mix and read in. assay protocol that is compatible with high throughput screening.
Catalog No. NC9945170