Learn More
AnaSpec SensoLyte Red Glucocerebrosidase (GBA) Assay Kit *Fluorimetric*
Catalog No. NC1739453
Encompass
The SensoLyte Red Glucocerebrosidase (GBA) Assay Kit detects GBA activity by using a highly sensitive fluorogenic substrate. The GBA substrate provided in the kit releases the red fluorescent dye resorufin upon Glucocerebrosidase cleavage with absorption/emission maxima at 570 nm/610 nm. This assay kit can be used to detect enzyme activity in purified enzyme preparations and compound screening. The Red Glucocerebrosidase (GBA) Assay is one-step homogenous reaction, which does not require the additional step of dispensing stop solution after the incubation, thereby suitable for HTS. The long wavelength fluorescence resorufin is also less interfered by the autofluorescence of components in biological samples and test compounds.
Catalog No. NC1739453 Supplier AnaSpec Supplier No. AS72259
The SensoLyte Red Glucocerebrosidase (GBA) Assay Kit detects GBA activity by using a highly sensitive fluorogenic substrate. The GBA substrate provided in the kit releases the red fluorescent dye resorufin upon Glucocerebrosidase cleavage with absorption/emission maxima at 570 nm/610 nm. This assay kit can be used to detect enzyme activity in purified enzyme preparations and compound screening. The Red Glucocerebrosidase (GBA) Assay is one-step homogenous reaction, which does not require the additional step of dispensing stop solution after the incubation, thereby suitable for HTS. The long wavelength fluorescence resorufin is also less interfered by the autofluorescence of components in biological samples and test compounds.