Learn More
AnaSpec SENSOLYTE RH110 MATRIPTASE
Supplier:  AnaSpec AS72241
Encompass
SensoLyte Rh110 Matriptase Activity Assay Kit Fluorimetric, (1 kit) Matriptase (also known as MT-SP1, ST14, TADG-15 and epithin) is a trypsin-like protease from the family of type II transmembrane serine proteases. The spectrum of known Matriptase substrates includes extracellular matrix proteins, cell adhesion molecules, ion channels, growth-factor-like proteins and other proteases. Its actions can result in protein processing, activation or degradation. Matriptase is widely expressed in virtually all epithelium and is specifically found in tumors of epithelial origin. Matriptase has been implicated in carcinogenesis including ovarian, prostate and cervical cancers and is considered a drug target for developing cancer therapeutic agent (1-3). This kit provides a convenient assay for screening of enzyme inhibitors and activators or for continuous assay of enzyme activity using a fluorogenic substrate. (excitation/emission=496 nm/520 nm)
Catalog No. NC1044751