Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One SensoPlate™ 24 Well Flat Bottomed Microplates with Lid
GREENER_CHOICE

Catalog No. 07001013 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-001-013 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-001-013 Supplier Greiner Bio-One Supplier No. 662892P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allows transmission measurements in 350nm range.

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Optically clear borosilicate glass bottom with light path of 175±15μm, black polystyrene me

Applications:

  • Fluorescence correl

Order Info

Comes as one each per case.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Array 4 x 6
Color Black
Material Polystyrene
No. of Wells 24
Sterility Sterile
Well Shape Flat Bottom (F-well)
Well Design Flattened, raised ring, Chimney
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.