Learn More
Greiner Bio-One SensoPlate™ 96-well, Non-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Catalog No. 07000633 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Catalog No. Color
07-000-633 Black, Clear Bottom
1 options
Catalog No. 07-000-633 Supplier Greiner Bio-One Supplier No. 655896
Add to Cart
Add to Cart
Color:Black, Clear Bottom; Sterility:Sterile; Lid:Without Lid
  • Round well geometry to reduce optical distortions
  • Black/White frame upon which 175μm borosilicate glass bottom is glued
  • Chimney-well prevents cross-contamination
  • Available with or without lid
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
Quantity 16/Cs.
Well Color Clear Bottom
Well Design Chimney