Learn More
Greiner Bio-One SensoPlate™ Glass Bottom Microplates
Catalog No. 07001043 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
07-001-043 Black
1 options
Catalog No. 07-001-043 Supplier Greiner Bio-One Supplier No. 781892P
Add to Cart
Add to Cart
For fluorescence correlation spectroscopy.
  • Low autofluorescence and high optical clarity
  • Borosilicate glass bottom and black polystyrene frame
  • Biocompatible adhesive
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Description Biocompatible adhesive
Array 16 x 24
Color Black
Material Glass
No. of Wells 384
Sterility Sterile
Well Shape Flat Bottom (F-well)