Learn More
Greiner Bio-One SensoPlate™ LoBase Glass-Bottom 1536-well Plate
GREENER_CHOICE
Catalog No. 07000020 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-020 Clear
1 options
Catalog No. 07-000-020 Supplier Greiner Bio-One Supplier No. 783892
Add to Cart
Add to Cart
Compatible with cell culture media
Alphanumeric well coding
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Array 32 x 48
Autoclavable Not Autoclavable
Color Clear
Material Borosilicate Glass, Polystyrene
No. of Wells 1536
Volume (Metric) Working 3 to 10 μL
Sterility Sterile
Well Shape Rounded Square
No. per Case 16
No. per Pack 1
Quantity 16/Cs.
Temperature Range (Metric) 4°C to 37°C
Dimensions (L x W x H) 128 x 85 x 10 mm