Learn More
Greiner Bio-One SensoPlate™ Plus Glass Bottom Microplates
GREENER_CHOICE
Catalog No. 07001041 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
07-001-041 Black
1 options
Catalog No. 07-001-041 Supplier Greiner Bio-One Supplier No. 781856P
Add to Cart
Add to Cart
For high-content screening applications.
  • Enables microscopy over surface of bottom of microplate
  • Flat-bottom well profile
  • 384 wells
  • Black
  • Ultra low base
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Description Ultra low base
Array 16 x 24
Color Black
Product Type Glass-bottom Microplate
Material Glass
No. of Wells 384
Well Shape Flat Bottom (F-well)