Learn More
World Precision Instrument Sensor, 150-380G
SDP
Catalog No. 502538757
Encompass
The tail cuff sensor has the ability to detect mouse/rat tail cuff blood pressure via the tail photoelectrically. Sensors are nonmagnetic and extremely sensitive in detecting pulses with the use of the II-MRBP systems.
Catalog No. 50-253-8757 Supplier World Precision Instrument Supplier No. IIB60044
The tail cuff sensor has the ability to detect mouse/rat tail cuff blood pressure via the tail photoelectrically. Sensors are nonmagnetic and extremely sensitive in detecting pulses with the use of the II-MRBP systems.