Learn More
World Precision Instrument Sensor, 20-80g
SDP
Catalog No. 502538759
Encompass
  • The tail cuff sensor has the ability to detect mouse/rat tail cuff blood pressure via the tail photoelectrically.
  • All the sensors are nonmagnetic.
  • They are extremely sensitive in detecting pulses with the use of the II-MRBP systems.
  • The ambient temperature that a reading can be taken starts at 30 degrees C. 
  • The systolic readings are within five mmHg of Cannulation and Telemetry. (Validated)
Catalog No. 50-253-8759 Supplier World Precision Instrument Supplier No. IIB60025
  • The tail cuff sensor has the ability to detect mouse/rat tail cuff blood pressure via the tail photoelectrically.
  • All the sensors are nonmagnetic.
  • They are extremely sensitive in detecting pulses with the use of the II-MRBP systems.
  • The ambient temperature that a reading can be taken starts at 30 degrees C. 
  • The systolic readings are within five mmHg of Cannulation and Telemetry. (Validated)