Learn More
World Precision Instrument Sensor, 70-160G
SDP
Catalog No. 502538758
Encompass
The tail cuff sensor has the ability to detect mouse/rat tail cuff blood pressure via the tail photoelectrically. All the sensors are nonmagnetic, and extremely sensitive in detecting pulses with the use of the II-MRBP systems.
Catalog No. 50-253-8758 Supplier World Precision Instrument Supplier No. IIB60038
The tail cuff sensor has the ability to detect mouse/rat tail cuff blood pressure via the tail photoelectrically. All the sensors are nonmagnetic, and extremely sensitive in detecting pulses with the use of the II-MRBP systems.