Learn More
Caron Products And Services Carbon Dioxide Sensor, 0-20%, Outputs 0-5 V
Catalog No. NC2596099
Encompass_Preferred
Carbon Dioxide Sensor, 0-20%, Outputs 0-5 V
Catalog No. NC2596099 Supplier Caron Products And Services Supplier No. CO2103
Carbon Dioxide Sensor, 0-20%, Outputs 0-5 V