Learn More
Mettler Toledo™ Sensor Holder, EasyPlace, Single Extended
Catalog No. 01911938 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
EasyPlace
Catalog No. For Use With (Equipment)
01-911-938 EasyPlace
1 options
This item is not returnable. View return policy
Catalog No. 01-911-938 Supplier Mettler Toledo™ Supplier No. 30671576
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sensor holder for the EasyPlace electrode arm with extended length for use with large sample vessels. Fits one sensor.
  • Extended Reach
  • For wide sample containers or elaborate experimental setups - the extended electrode holder provides additional horizontal reach
TRUSTED_SUSTAINABILITY
Specifications
Type Sensor Holder, EasyPlace Single Extended
For Use With (Equipment) EasyPlace