Learn More
Mettler Toledo™ Sensor Holder, EasyPlace, Triple
Catalog No. 01911937 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
EasyPlace
Catalog No. For Use With (Equipment)
01-911-937 EasyPlace
1 options
This item is not returnable. View return policy
Catalog No. 01-911-937 Supplier Mettler Toledo™ Supplier No. 30671575
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sensor holder for the EasyPlace electrode arm. Fits up to three sensors.
  • Three-in-One
  • For multi-channel measurements, external references or temperature sensors - position multiple sensor with one EasyPlace
TRUSTED_SUSTAINABILITY
Specifications
Type Sensor Holder, EasyPlace Triple
For Use With (Equipment) EasyPlace