Description
- Supports measurements in large containers approximately 5 to 15 L in volume.
- Designed for use with the EasyPlace Sensor Stand.
- Enables measurements even when sample fill levels are low.
Specifications
Specifications
| For Use With (Equipment) | pH Electrode |
| Type | Electrode Holder |