Learn More
Ace Glass, Inc. Sensor, J, 1/4x36in, inconel sheath, 12 foot lead
SDP
Catalog No. 50983334
Encompass
Sensor, J, 1/4x36in, inconel sheath, 12 foot lead
Catalog No. 50-983-334 Supplier Ace Glass, Inc. Supplier No. 1214014
Sensor, J, 1/4x36in, inconel sheath, 12 foot lead