Learn More
Hach Company Analog pH/ORP/Dissolved Oxygen Sensor Input Module for SC200 Universal Controller
Catalog No. 501997281 Shop All Hach Company Products
Encompass
The pH/ORP/Dissolved Oxygen module for the SC200 is a user installed module that allows for connection of analog pH, ORP, and dissolved oxygen sensors to the controller. This module works with combination and differential pH and ORP sensors as well as amperometric dissolved oxygen sensors. See manual for additional connection information.
Catalog No. 50-199-7281 Supplier Hach Company Supplier No. 9012900
The pH/ORP/Dissolved Oxygen module for the SC200 is a user installed module that allows for connection of analog pH, ORP, and dissolved oxygen sensors to the controller. This module works with combination and differential pH and ORP sensors as well as amperometric dissolved oxygen sensors. See manual for additional connection information.