Learn More
MSC Bradley Sensored Soap Dispenser, Lotion, Counter Mount, Chrome Plated
Catalog No. 502839823
Encompass
The Bradley 6315-KT0000 is a sensored soap dispenser starter kit designed for counter mounting. It features a chrome-plated plastic construction and dispenses lotion soap. The kit includes the soap dispenser, a 1000-shot bottle of soap, and four "D"-cell alkaline batteries, making it ready for immediate use. It is ADA compliant.
  • Sensored operation for touch-free use.
  • LED indicator lights for low battery and low soap refill.
  • Deck mounted design for easy installation on countertops.
  • Constructed from durable chrome-plated plastic.
  • ADA compliant for accessibility.
  • Starter kit includes a 1000-shot bottle of lotion soap and four "D"-cell batteries.
  • Dispenses 0.027 oz of lotion soap with moisturizer per activation.
Catalog No. 50-283-9823 Supplier MSC Supplier No. 94906377
The Bradley 6315-KT0000 is a sensored soap dispenser starter kit designed for counter mounting. It features a chrome-plated plastic construction and dispenses lotion soap. The kit includes the soap dispenser, a 1000-shot bottle of soap, and four "D"-cell alkaline batteries, making it ready for immediate use. It is ADA compliant.
  • Sensored operation for touch-free use.
  • LED indicator lights for low battery and low soap refill.
  • Deck mounted design for easy installation on countertops.
  • Constructed from durable chrome-plated plastic.
  • ADA compliant for accessibility.
  • Starter kit includes a 1000-shot bottle of lotion soap and four "D"-cell batteries.
  • Dispenses 0.027 oz of lotion soap with moisturizer per activation.