Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILC Dover Sentinel XL Blower Assembly

Catalog No. 19631094
Change view
Click to view available options
For Use With (Equipment):
Sentinel XL HP System and CBRN PAPR
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-631-094 Sentinel XL HP System and CBRN PAPR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-631-094 Supplier ILC Dover Supplier No. S2002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Sentinel XL Blower has a rugged, sealed nylon protective shell, with floating polycarbonate internal blower assembly.

  • Dual chamber provides enhanced durability
  • Can be used with alkaline battery pack or a NiMH rechargeable battery

Specifications

Color Gray
Product Type Motor Blower
For Use With (Equipment) Sentinel XL HP System and CBRN PAPR
Model S-2002
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.