Learn More
ILC Dover Sentinel XL HP Clear Hood System with Duffel
Catalog No. 19631072
Change view
Click to view available options
Includes: Duffel Bag
Catalog No.
19-631-072 Includes: Duffel Bag
1 options
Catalog No. 19-631-072 Supplier ILC Dover Supplier No. S300016
Add to Cart
Add to Cart
Ideal for healthcare settings, the Sentinel XL HP Clear Hood System includes S-3101 hood, breathing tube, XL blower, flow meter, plug, NiMH rechargeable battery, charging cord, quick loc belt, 2 HEPA filters, and carrying tote.
  • Ideal for use in Healthcare settings
  • Sentinel XL powered air-purifying respirator assemblies and systems include parts required to build a full system or to replace existing parts
  • Belt-mounted blower distributes ambient air through a filter to provide clean, breathable air to the wearer
  • PAPRs allow workers to move freely and unrestricted
  • Sentinel XL system parts are compatible with components of the same brand and series
Specifications
Product Type PAPR System with Hood
Model Sentinel XL