Learn More
ILC Dover Sentinel XL HP Half Hood
Catalog No. 19631098
Change view
Click to view available options
Quantity: 25/Pk.
Catalog No.
19-631-098 Quantity: 25/Pk.
1 options
Catalog No. 19-631-098 Supplier ILC Dover Supplier No. S400125
Add to Cart
Add to Cart
Half hood head cover offers an APF of 25, provides a wide field of view, and easily accommodates stethoscopes.
  • Offers an APF of 25
  • Provides a wide field of view and easily accommodates stethoscopes
Specifications
Product Type Half Hood
Model S-4001-25
Quantity 25/Pk.