Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILC Dover Sentinel XT Blower

Catalog No. 19631105
Catalog No. 19-631-105 Supplier ILC Dover Supplier No. S3114
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Surprisingly lightweight design when paired with ILC Dover's lithium polymer battery. Blower produces 8+ CFM airflow, exceeding NIOSH requirements.

  • Brushless motor design for long life and continued quiet operation
  • Secondary impact protection chamber for air handling unit
  • Approved by Factory Mutual for use in Class I, II, and III; Division 2; Groups C, D, E, F and G

Specifications

Certifications/Compliance Factory Mutual for use in Class I, II, and III; Division 2; Groups C, D, E, F and G
Color Gray
Product Type Motor Blower
For Use With (Equipment) Sentinel PAPR System
Model S-3114
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.