Learn More
ILC Dover Sentinel XT Clear Hoods
Catalog No. 19631106
Change view
Click to view available options
Quantity: 30/Pk.
Catalog No.
19-631-106 Quantity: 30/Pk.
1 options
Catalog No. 19-631-106 Supplier ILC Dover Supplier No. S311230
Add to Cart
Add to Cart
Sentinel XT clear hood for use with the Sentinel XT Blower.
  • With an APF of 1,000, it offers a full 320-degree field of view, allows the wearer to have facial hair and/or a head covering, and reduces feelings of claustrophobia
  • Metal clips allow the outer bib to secure to protective clothing
Specifications
Product Type Respirator Hood
Model S-3112-30
Quantity 30/Pk.