Learn More
ILC Dover Sentinel XT Clear Plus Hood
Catalog No. 19631082
Change view
Click to view available options
Quantity: 30/Pk.
Catalog No.
19-631-082 Quantity: 30/Pk.
1 options
Catalog No. 19-631-082 Supplier ILC Dover Supplier No. S332430
Add to Cart
Add to Cart
Sentinel XT clear plus hood for use with the Sentinel XT Blower.
  • With an APF of 1,000, it offers a full 320-degree field of view, allows the wearer to have facial hair and/or a head covering, and reduces feelings of claustrophobia
  • Wrap around straps allow the outer bib to secure to protective clothing
Specifications
For Use With (Equipment) Sentinel XT Blower
Model S-3324-30
Quantity 30/Pk.